Anti-MAP3K6 Rabbit Polyclonal Antibody
Catalog # ABGEAP7912A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase Kinase Kinase 6
- Antigen symbol:MAP3K6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal (765–781 aa)
- Cross adsorption:No
- Gene ID:389856
- Antigen synonyms:ASK2, MAPKKK6, MEKK6
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:YDKDIEQIAKEEQGC
- Storage temperature:Cool
- Concentration:0,35 mg/ml
- Shipping temperature:Ice
- Immunogen:This USP27X antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human USP27X.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MAP3K6 (mitogen-activated protein kinase kinase kinase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse