Anti-CA3 Rabbit Polyclonal Antibody
Catalog # ABGEAP7633A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 3
- Antigen symbol:CA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:CAIII|Car3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CA3 (carbonic anhydrase III, muscle specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse