Anti-ERBB4 Rabbit Polyclonal Antibody
ABGEAP7631B
: Abgent
- Antibody type:Primary
- Antigen name:Erb-b2 receptor tyrosine kinase 4
- Antigen symbol:ERBB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (1291–1308 aa)
- Cross adsorption:No
- Antigen synonyms:ERBB4|Verb a e|Tyrosine kinase type cell surface receptor HER4|p180erbB4|Avian erythroblastic leukemia viral (verbb2) oncogene homolog 4|C-erbB-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ERBB4 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human