Anti-ADAMTS5 Rabbit Polyclonal Antibody
ABGEAP7447C
: Abgent
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase with Thrombospondin Type 1 Motif 5
- Antigen symbol:ADAMTS5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:ADAMTS-11|ADAM-TS 5|ADAMTS-5|ADAMTS11|ADAM-TS5|ADAM-TS 11|ADMP-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ADAMTS5 (ADAM metallopeptidase with thrombospondin type 1 motif, 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse