Order Entry
Italy
ContactUsLinkComponent
 
Anti-SPRY4 Rabbit Polyclonal Antibody
  ABGEAP7446B
 :  Abgent
undefined
Anti-SPRY4 Rabbit Polyclonal Antibody
  ABGEAP7446B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Sprouty RTK Signaling Antagonist 4
  • Antigen symbol:
    SPRY4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    Spry-4|Protein sprouty homolog 4|SPRY4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: SPRY4 (sprouty homolog 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human