Order Entry
Italy
ContactUsLinkComponent
Anti-COL4A1 Rabbit Polyclonal Antibody
Anti-COL4A1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7369A
Supplier:  Abgent
undefined
Anti-COL4A1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7369A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    collagen, type IV, alpha 1
  • Antigen symbol:
    COL4A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Antigen synonyms:
    FREAC7|FKHL11|FKH6
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: COL4A1 (collagen, type IV, alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse