Anti-C1QA Rabbit Polyclonal Antibody
Catalog # ABGEAP7324B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Complement Component 1, q subcomponent
- Antigen symbol:C1QA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:A chain|q subcomponent|complement C1q subcomponent subunit A|complement component 1|complement component C1q|alpha polypeptide
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: C1QA (complement component 1, q subcomponent, A chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human