Order Entry
Italy
ContactUsLinkComponent
Anti-MAPK10 Rabbit Polyclonal Antibody
Anti-MAPK10 Rabbit Polyclonal Antibody
Catalog # ABGEAP7222A
Supplier:  Abgent
undefined
Anti-MAPK10 Rabbit Polyclonal Antibody
Catalog # ABGEAP7222A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 10
  • Antigen symbol:
    MAPK10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal (1–100 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    MAPK10|JNK3|JNK3A|JNK 46|MGC50974|Mitogen activated protein kinase 10|JNK1A2|MAP kinase p49 3F12|MAPK 8|JNK|MAP kinase 9|MAPK9|JNK 55|JNK2|Mitogen activated protein kinase 9|MAP kinase 8|JNK2BETA|JNK1|p54bSAPK|PRKM10|JNK2B|Mitogen activated protein kinase 8|MAPK8|JNK2ALPHA|JNK21B1/2|MAPK 9
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MAPK10 (mitogen-activated protein kinase 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse