Anti-NDRG1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6935A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:N-Myc Downstream Regulated 1
- Antigen symbol:NDRG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:TDD5|RIT42|HMSNL|PROXY1|CAP43,|protein regulated by oxygen-1|GC4|NDR1|reducing agents and tunicamycin-responsive protein|tunicamycin-responsive protein|nickel-specific induction protein Cap43|N-myc downstream regulated gene 1 protein|TARG1|N-myc downstream regulated gene 1|differentiation-related gene 1 protein|NMSL|CMT4D|RTP|DRG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: NDRG1 (N-myc downstream regulated 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human