Order Entry
Italy
Orders LinkContactUsLinkComponent
Anti-TP53 Rabbit Polyclonal Antibody

Anti-TP53 Rabbit Polyclonal Antibody

Codice catalogo ABGEAP6266D
Nome del fornitore:   Abgent
Anti-TP53 Rabbit Polyclonal Antibody
Codice catalogo ABGEAP6266D
Nome del fornitore:  Abgent
Anti-TP53 Rabbit Polyclonal Antibody
Anti-TP53 Rabbit Polyclonal Antibody
Price
365,00 €
Log in for availability
1 * 400 µl
Product Details & Documents

Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human

Documents

Specifications
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Flow cytometry:
    Yes
  • Antibody type:
    Primary
  • Storage temperature:
    Cool
  • Host:
    Rabbit
  • Shipping temperature:
    Ice
  • Antigen name:
    Tumor protein p53
  • Cat. No.:
    ABGEAP6266D
  • Reactivity:
    Human
  • Antigen synonyms:
    Tumor suppressor p53|Cellular tumor antigen p53|TP53|Antigen NY-CO-13|Phosphoprotein p53
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    P53
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.