Anti-MLLT3 Rabbit Polyclonal Antibody
Catalog # ABGEAP6190B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Myeloid/lymphoid or Mixed-lineage Leukemia (trithorax homolog Drosophila); translocated to 3
- Antigen symbol:MLLT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:Center V422
- Cross adsorption:No
- Antigen synonyms:Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein|AF9|YEATS3|ALL1 fused gene from chromosome 9 protein
- Sequence:V422
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MLLT3 (myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human