Anti-LRP6 Rabbit Polyclonal Antibody
Catalog # ABGEAP6158A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Low density lipoprotein receptor-related protein 6
- Antigen symbol:LRP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (1500–1575 aa)
- Cross adsorption:No
- Antigen synonyms:LDL receptor-related protein 6|ADCAD2
- Sequence:T1546
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: LRP6 (low density lipoprotein receptor-related protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human