Anti-OTUD4 Rabbit Polyclonal Antibody
Catalog # ABGEAP5997A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:OTU Domain Containing 4
- Antigen symbol:OTUD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:HSHIN1|KIAA1046|HIV-1-induced protein HIN-1|DUBA6|OTU domain-containing protein 4|HIN1|OTUD4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: OTUD4 (OTU deubiquitinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human