Anti-COQ7 Rabbit Polyclonal Antibody
Catalog # ABGEAP5880B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Engrailed 1
- Antigen symbol:COQ7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:Homeobox protein engrailed-1|Hu-En-1|HME1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: COQ7 (coenzyme Q7 homolog, ubiquinone (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human