Anti-LSM4 Rabbit Polyclonal Antibody
Catalog # ABGEAP5835C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:LSM4 Homolog, U6 Small Nuclear RNA And MRNA Degradation Associated
- Antigen symbol:LSM4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:GRP|YER112W
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: LSM4 (LSM4 homolog, U6 small nuclear RNA associated (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse