Anti-GP6 Rabbit Polyclonal Antibody
ABGEAP5691B
: Abgent
- Antibody type:Primary
- Antigen name:Glycoprotein 6
- Antigen symbol:GP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:platelet collagen receptor|GP6|gp 6|gp6|glycoprotein 6|Glycoprotein VI
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: GP6 (glycoprotein VI (platelet))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human