Anti-EID2 Rabbit Polyclonal Antibody
Catalog # ABGEAP5633B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:RGP1 Homolog, RAB6A GEF Complex Partner 1
- Antigen symbol:EID2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:KIAA0258|RGP1|Retrograde Golgi transport protein RGP1 homolog
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: EID2 (EP300 interacting inhibitor of differentiation 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse