Order Entry
Italy
ContactUsLinkComponent
Anti-SLC9A9 Rabbit Polyclonal Antibody
Anti-SLC9A9 Rabbit Polyclonal Antibody
Catalog # ABGEAP5412B
Supplier:  Abgent
undefined
Anti-SLC9A9 Rabbit Polyclonal Antibody
Catalog # ABGEAP5412B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 9 Member 9
  • Antigen symbol:
    Solute carrier family 9, member 9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    5730527A11Rik|AI854429|9930105B05|Nhe9
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SLC9A9 (solute carrier family 9, subfamily A (NHE9, cation proton antiporter 9), member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse