Anti-BHLHE22 Rabbit Polyclonal Antibody
Catalog # ABGEAP4777C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Basic Helix-Loop-Helix Family Member E22
- Antigen symbol:BHLHE22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:GPR126|Vascular inducible G protein-coupled receptor|G-protein coupled receptor 126|Developmentally regulated G-protein-coupled receptor|PS1TP2|DREG|VIGR|APG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BHLHE22 (basic helix-loop-helix family, member e22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human