Anti-GIT1 Rabbit Polyclonal Antibody
ABGEAP3553A
: Abgent
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor Kinase Interacting ArfGAP 1
- Antigen symbol:GIT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:GIT1|ARF GTPase-activating protein GIT1|CAT1|G protein-coupled receptor kinase-interactor 1|GRK-interacting protein 1|CAT-1|ARF GAP GIT1|Cool-associated and tyrosine-phosphorylated protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: GIT1 (G protein-coupled receptor kinase interacting ArfGAP 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human