Order Entry
Italy
ContactUsLinkComponent
Anti-MEF2C Rabbit Polyclonal Antibody
Anti-MEF2C Rabbit Polyclonal Antibody
Catalog # ABGEAP3382A
Supplier:  Abgent
undefined
Anti-MEF2C Rabbit Polyclonal Antibody
Catalog # ABGEAP3382A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myocyte enhancer factor 2C
  • Antigen symbol:
    MEF2C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    MADS box transcription enhancer factor 2 polypeptide C myocyte enhancer factor 2C|Myocyte enhancer factor 2C|Similar to MADS box transcription enhancer factor 2 polypeptide C|MEF2Cphospho S387|MEF2C_HUMAN.|Myocyte specic enhancer factor 2C
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human