Anti-TP53 Rabbit Polyclonal Antibody
Catalog # ABGEAP3200A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Tumor protein p53
- Antigen symbol:P53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:Tumor suppressor p53|Cellular tumor antigen p53|TP53|Antigen NY-CO-13|Phosphoprotein p53
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human