Order Entry
Italy
ContactUsLinkComponent
Anti-NMRK2 Rabbit Polyclonal Antibody
Anti-NMRK2 Rabbit Polyclonal Antibody
Catalog # ABGEAP2791A
Supplier:  Abgent
undefined
Anti-NMRK2 Rabbit Polyclonal Antibody
Catalog # ABGEAP2791A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nicotinamide riboside kinase 2
  • Antigen symbol:
    NMRK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    Muscle integrin-binding protein|NmR-K 2|MIBP|Integrin beta-1-binding protein 3|Nicotinamide riboside kinase 2|Ribosylnicotinamide kinase 2|NRK 2|Ribosylnicotinic acid kinase 2|RNK 2|NRK2|ITGB1BP3|Nicotinic acid riboside kinase 2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: NMRK2 (nicotinamide riboside kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human