Order Entry
Italy
ContactUsLinkComponent
Anti-LGR5 Rabbit Polyclonal Antibody
Anti-LGR5 Rabbit Polyclonal Antibody
Catalog # ABGEAP2745D
Supplier:  Abgent
Anti-LGR5 Rabbit Polyclonal Antibody
Catalog # ABGEAP2745D
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Leucine-rich Repeat Containing G protein-coupled receptor 5
  • Antigen symbol:
    LGR5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Loop 2
  • Antigen synonyms:
    GPR67|GPR49|G-protein coupled receptor 49|LGR5|G-protein coupled receptor HG38|G-protein coupled receptor 67|Leucine-rich repeat-containing G-protein coupled receptor 5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: LGR5/GPR49 (loop2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Loop 2
Host: Rabbit
Isotype:
Reactivity: Human, Mouse