Anti-SEMA6A Rabbit Polyclonal Antibody
Catalog # ABGEAP2740A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 6A
- Antigen symbol:SEMA6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:HT018|Sema Domain|SEMAQ|SEMA|(semaphorin) 6A|VIA|SEMA6A1|And cytoplasmic Domain|transmembrane Domain (TM)
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SEMA6A (sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human