Order Entry
Italy
ContactUsLinkComponent
Anti-RNF36 (TRIM69) Rabbit Polyclonal Antibody
Anti-RNF36 (TRIM69) Rabbit Polyclonal Antibody
  ABGEAP2525D
 :  Abgent
undefined
Anti-RNF36 (TRIM69) Rabbit Polyclonal Antibody
  ABGEAP2525D
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Tripartite Motif Containing 69
  • Antigen symbol:
    TRIM69
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    POP2
  • Sequence:
    K251
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: RNF36 (TRIM69)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human