Anti-RNF36 (TRIM69) Rabbit Polyclonal Antibody
Catalog # ABGEAP2525C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 69
- Antigen symbol:TRIM69
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:POP2
- Sequence:H215
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RNF36 (TRIM69)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human