Anti-TPST2 Rabbit Polyclonal Antibody
Catalog # ABGEAP2322B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Tyrosylprotein Sulfotransferase 2
- Antigen symbol:TPST2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:Protein-tyrosine sulfotransferase 2|Protein tyrosine sulfotransferase 2|EC 2.8.2.20.|TPST2_HUMAN|TPST-2|Tpst2|Tyrosylprotein phosphotransferase 2|Tyrosylprotein sulfotransferase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TPST2 (tyrosylprotein sulfotransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human