Order Entry
Italy
ContactUsLinkComponent
Anti-RBCK1 (UBCE7IP3) Rabbit Polyclonal Antibody
Anti-RBCK1 (UBCE7IP3) Rabbit Polyclonal Antibody
Catalog # ABGEAP2107A
Supplier:  Abgent
undefined
Anti-RBCK1 (UBCE7IP3) Rabbit Polyclonal Antibody
Catalog # ABGEAP2107A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RanBP-type and C3HC4-type zinc finger containing 1
  • Antigen symbol:
    RBCK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal ( 1–50 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    XAP4|PBMEI|Ubiquitin conjugating enzyme 7 interacting protein 3|Hepatitis B virus X-associated protein 4|HBV associated factor 4|XAP3|HOIL-1|RBCK2|UBCE7IP3|HOIL1|ZRANB4|PGBM1|RNF54|C20orf18
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: RBCK1 (UBCE7IP3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse