Anti-RMI1 Rabbit Polyclonal Antibody
ABGEAP16875A
: Abgent
- Antibody type:Primary
- Antigen name:RecQ mediated genome instability 1
- Antigen symbol:RMI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:80010
- Antigen synonyms:C9orf76|BLAP75|FAAP75
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:70.144 kDa
- Sequence:AQVTPKPWEAKPSRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This RMI1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 105-133 amino acids from the N-terminal region of human RMI1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: RMI1 (RecQ mediated genome instability 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human