Anti-SNX2 Rabbit Polyclonal Antibody
Catalog # ABGEAP16134B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Sorting Nexin 2
- Antigen symbol:SNX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:6643
- Antigen synonyms:MGC5204|TRG-9
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:58.471 kDa
- Sequence:RKEVGRFEKERVKDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SNX2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 457-485 amino acids from the C-terminal region of human SNX2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SNX2 (sorting nexin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human