Anti-PTK2 Rabbit Polyclonal Antibody
ABGEAP14458C
: Abgent
- Antibody type:Primary
- Antigen name:Protein-tyrosine Kinase 2
- Antigen symbol:PTK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitope:Center
- Gene ID:14083
- Antigen synonyms:FAK-related non-kinase polypeptide|protein-tyrosine kinase 2|Focal Adhesion Kinase|FADK 1|focal adhesion kinase 1
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:123.537 kDa
- Sequence:PSKSYGIDEARDYC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Ptk2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 426-453 amino acids from the Central region of mouse Ptk2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse