Anti-MST1R Rabbit Polyclonal Antibody
ABGEAP14137B
: Abgent
- Antibody type:Primary
- Antigen name:Macrophage Stimulating 1 Receptor
- Antigen symbol:MST1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:soluble R|RON variant E2E3|MST1R variant RON30|PTK8|macrophage-stimulating protein receptor|MST1R variant RON62|soluble RON variant 1|c-met-related tyrosine kinase|CDw136|PTK8 protein tyrosine kinase 8|MSP receptor|p185-Ron|soluble RON variant 3|RON|soluble RON variant 2|CD136
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: MST1R (macrophage stimulating 1 receptor (c-met-related tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse