Order Entry
Italy
ContactUsLinkComponent
Anti-CSNK1G1 Rabbit Polyclonal Antibody
Anti-CSNK1G1 Rabbit Polyclonal Antibody
Catalog # ABGEAP14134B
Supplier:  Abgent
undefined
Anti-CSNK1G1 Rabbit Polyclonal Antibody
Catalog # ABGEAP14134B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Casein kinase 1, gamma 1
  • Antigen symbol:
    CSNK1G1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    Casein kinase I isoform gamma-1|CK-I gamma1|CKI gamma 1|CSNK1G1|CK1G1|CK I gamma1|CKI- gamma1|CKI gamma-1|KC1G1|CKI-gamma-1|CKI-gamma 1|CKI gamma1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CSNK1G1 (casein kinase 1, gamma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse