Anti-IBSP Rabbit Polyclonal Antibody
ABGEAP14114A
: Abgent
- Antibody type:Primary
- Antigen name:Integrin-Binding Sialoprotein
- Antigen symbol:IBSP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:BSP-II|BNSP|SP-II|BSP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: IBSP (integrin-binding sialoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human