Order Entry
Italy
ContactUsLinkComponent
Anti-DCBLD2 Rabbit Polyclonal Antibody
Anti-DCBLD2 Rabbit Polyclonal Antibody
  ABGEAP13827B
 :  Abgent
undefined
Anti-DCBLD2 Rabbit Polyclonal Antibody
  ABGEAP13827B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Discoidin, CUB and LCCL domain containing 2
  • Antigen symbol:
    DCBLD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    ESDN|coagulation factor V/VIII-homology domains protein 1|endothelial and smooth muscle cell-derived neuropilin-like protein|CLCP1|DCBD2_HUMAN.|LCCL and coagulation factor V/VIII-homology domains protein 1|Discoidin|CUB and LCCL domain-containing protein 2|CUB
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: DCBLD2 (discoidin, CUB and LCCL domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse