Order Entry
Italy
ContactUsLinkComponent
Anti-SLC2A13 Rabbit Polyclonal Antibody
Anti-SLC2A13 Rabbit Polyclonal Antibody
Catalog # ABGEAP13677C
Supplier:  Abgent
undefined
Anti-SLC2A13 Rabbit Polyclonal Antibody
Catalog # ABGEAP13677C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 2 Member A13
  • Antigen symbol:
    Solute carrier family 2, member 13
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    FREAC8|FKHL12
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SLC2A13 (solute carrier family 2 (facilitated glucose transporter), member 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse