Anti-SLC30A6 Rabbit Polyclonal Antibody
Catalog # ABGEAP13600C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 30 Member A6
- Antigen symbol:Solute carrier family 30, member 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:MSTP103|solute carrier family 30 zinc transporter member 6|ZNT6|MST103
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SLC30A6 (solute carrier family 30 (zinc transporter), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse