Anti-SIDT2 Rabbit Polyclonal Antibody
ABGEAP13596C
: Abgent
- Antibody type:Primary
- Antigen name:SID1 transmembrane family, member 2
- Antigen symbol:SIDT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:CGI-40|SIDT2|SID1 transmembrane family member 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: SIDT2 (SID1 transmembrane family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse