Anti-NDUFAF2 Rabbit Polyclonal Antibody
ABGEAP13341C
: Abgent
- Antibody type:Primary
- Antigen name:NADH dehydrogenase (ubiquinone) complex 1, assembly factor 2
- Antigen symbol:NDUFAF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center
- Gene ID:91942
- Antigen synonyms:Mimitin|Myc-induced mitochondrial protein|B17.2L|mitochondrial|mimitin|MMTN|Mimitin Antibody|NDUFA12L
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:19.856 kDa
- Sequence:EEILKNEKHREEIKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NDUFAF2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 71-99 amino acids from the Central region of human NDUFAF2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: NDUFAF2 (NADH dehydrogenase (ubiquinone) complex I, assembly factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human