Anti-BNIP3 (NIP3) Rabbit Polyclonal Antibody
Catalog # ABGEAP1321A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:BCL2/adenovirus E1B 19-kDa protein-interacting protein 3
- Antigen symbol:BNIP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:1–100 aa
- Cross adsorption:No
- Antigen synonyms:NIP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BNIP3 (NIP3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: BH3 domain
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse