Anti-HSD17B4 Rabbit Polyclonal Antibody
Catalog # ABGEAP12516C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid (17-beta) Dehydrogenase 4
- Antigen symbol:HSD17B4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:MFE-2|PRLTS1|DBP|SDR8C1|MPF-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: HSD17B4 (hydroxysteroid (17-beta) dehydrogenase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human