Order Entry
Italy
ContactUsLinkComponent
Anti-MAP3K15 Rabbit Polyclonal Antibody
Anti-MAP3K15 Rabbit Polyclonal Antibody
Catalog # ABGEAP12323C
Supplier:  Abgent
undefined
Anti-MAP3K15 Rabbit Polyclonal Antibody
Catalog # ABGEAP12323C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase Kinase Kinase 15
  • Antigen symbol:
    MAP3K15
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    ASK3, bA723P2.3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MAP3K15 (mitogen-activated protein kinase kinase kinase 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse