Anti-SET7 (SET9) Rabbit Polyclonal Antibody
ABGEAP1194C
: Abgent
- Antibody type:Primary
- Antigen name:SET Domain Containing (lysine methyltransferase) 7 And 9
- Antigen symbol:SET7 / SET9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:Center (199–214 aa)
- Cross adsorption:No
- Antigen synonyms:SET9
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: SET7 (SET9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse