Order Entry
Italy
ContactUsLinkComponent
 
Anti-PTPN20A Rabbit Polyclonal Antibody
  ABGEAP11798C
 :  Abgent
Anti-PTPN20A Rabbit Polyclonal Antibody
  ABGEAP11798C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 46, member A
  • Antigen symbol:
    PTPN20A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    XTP11|C6orf37
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: PTPN20A (protein tyrosine phosphatase, non-receptor type 20A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human