Anti-OR4F15 Rabbit Polyclonal Antibody [clone: 199CT6.1.2]
Catalog # ABGEAP11344A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Olfactory Receptor, Family 4, Subfamily F Member 15
- Antigen symbol:OR4F15
- Clonality:Polyclonal
- Clone:199CT6.1.2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:Endogenous carboxypeptidase inhibitor|ECI|Latexin protein|TCI|MUM|Tissue carboxypeptidase inhibitor.|Latexin
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: OR4F15 (olfactory receptor, family 4, subfamily F, member 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human