Anti-NXPH4 Rabbit Polyclonal Antibody
ABGEAP11180B
: Abgent
- Antibody type:Primary
- Antigen name:Neurexophilin 4
- Antigen symbol:NXPH4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Antigen synonyms:UNQ1928/PRO4403|Neurexophilin 4|NPH4|NXPH4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: NXPH4 (neurexophilin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human