Anti-GPAA1 Rabbit Polyclonal Antibody
Catalog # ABGEAP10722A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Glycosylphosphatidylinositol anchor attachment Protein 1 homolog (yeast)
- Antigen symbol:GPAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal (1–100 aa)
- Cross adsorption:No
- Antigen synonyms:hGAA1|GAA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: GPAA1 (glycosylphosphatidylinositol anchor attachment 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human