Order Entry
Italy
ContactUsLinkComponent
Anti-MPP1 Rabbit Polyclonal Antibody
Anti-MPP1 Rabbit Polyclonal Antibody
Catalog # ABGEAP10708C
Supplier:  Abgent
Anti-MPP1 Rabbit Polyclonal Antibody
Catalog # ABGEAP10708C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Membrane Palmitoylated Protein 1
  • Antigen symbol:
    MPP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Antigen synonyms:
    DXS552E|AAG12|MRG1|55 kDa erythrocyte membrane protein|PEMP|EMP55
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MPP1 (membrane protein, palmitoylated 1, 55kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human