Order Entry
Italy
ContactUsLinkComponent
Anti-TLE4 Mouse Monoclonal Antibody
Anti-TLE4 Mouse Monoclonal Antibody
Catalog # ABGEAM2017A
Supplier:  Abgent
undefined
Anti-TLE4 Mouse Monoclonal Antibody
Catalog # ABGEAM2017A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Transducin-like enhancer of split 4
  • Antigen symbol:
    TLE4
  • Clonality:
    Monoclonal
  • Clone:
    384CT24.1.3.4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Center
  • Cross adsorption:
    No
  • Form:
    Ascites
  • Gene ID:
    7091
  • Antigen synonyms:
    ESG4|Grg-4|ESG|BCE1|GRG4|BCE-1|E(spI)
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    83.755 kDa
  • Sequence:
    NGLDKTRLLKKDAPC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Shipping temperature:
    4 °C
  • Immunogen:
    This TLE4 antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 262-290 amino acids from the Central region of human TLE4.
  • Size:
    400 µl
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: TLE4 (transducin-like enhancer of split 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Mouse
Isotype: none
Reactivity: Human